3-[1,2,4]Triazol-4-ylphenylamine ≥95%

Supplier: Ambeed
Danger

A517319-1G A517319-250MG
77882-268EA 103 USD
77882-268 77882-269
3-[1,2,4]Triazol-4-ylphenylamine ≥95%
3-[1,2,4]Triazol-4-ylphenylamine

Precaución: for Research Use Only

Formula: C₈H₈N₄
MDL Number: MFCD07395923
CAS Number: 252928-92-0
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR