Potassium(iodomethyl)trifluoroborate ≥96%

Supplier: Ambeed
Warning

Synonyms: Potassium trifluoro(iodomethyl)borate

A516846-100MG A516846-1G A516846-250MG
77880-116EA 25 USD
77880-116 77880-117 77880-118
Potassium(iodomethyl)trifluoroborate ≥96%
Potassium (iodomethyl)trifluoroborate

Precaución: for Research Use Only

Formula: CH₂BF₃IK
MW: 247.83 g/mol
MDL Number: MFCD11112187
CAS Number: 888711-47-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR