(1-(tert-Butyl)-1H-pyrazol-4-yl)boronic acid 98+%, Ambeed.Inc
Supplier: Ambeed
|
Warning
|
A516730-100MG
A516730-250MG
A516730-1G
77684-343EA
33
USD
77684-343
77891-044
77891-043
(1-(tert-Butyl)-1H-pyrazol-4-yl)boronic acid 98+%, Ambeed.Inc
(1-(tert-Butyl)-1H-pyrazol-4-yl)boronic acid
Precaución: For research use only.
Formula:
C₇H₁₃BN₂O₂ |
MDL Number:
MFCD18250635 CAS Number: 1416785-99-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...