ABT-888 ≥99%

Supplier: Ambeed
Warning

Synonyms: (R)-2-(2-Methylpyrrolidin-2-yl)-1H-benzo[d]imidazole-7-carboxamide dihydrochloride, Veliparib 2HCl

A516702-10MG A516702-5MG A516702-100MG A516702-250MG A516702-50MG
77669-296EA 63 USD
77669-296 77669-298 77872-864 77872-865 77872-866
ABT-888 ≥99%
ABT-888

Precaución: for Research Use Only

Formula: C₁₃H₁₈Cl₂N₄O
MW: 317.214 g/mol
MDL Number: MFCD12407402
CAS Number: 912445-05-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR