(R)-tert-Butyl (1-hydroxybutan-2-yl)carbamate ≥98%
Supplier: Ambeed
|
Danger
|
A507826-25G
A507826-1G
A507826-5G
77686-325EA
237
USD
77686-325
77901-819
77901-257
(R)-tert-Butyl (1-hydroxybutan-2-yl)carbamate ≥98%
(R)-tert-Butyl (1-hydroxybutan-2-yl)carbamate
Precaución: for Research Use Only
Formula:
C₉H₁₉NO₃ |
MDL Number:
MFCD04974273 CAS Number: 150736-71-3 UN: 2923 ADR: 8,6.1,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...