2,4-Difluoro-1,3,5-trimethylbenzene 95+%

Supplier: Ambeed
Danger

A507709-100MG A507709-1G A507709-250MG
77749-034EA 20 USD
77749-034 77749-035 77749-036
2,4-Difluoro-1,3,5-trimethylbenzene 95+%
2,4-Difluoro-1,3,5-trimethylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₀F₂
MW: 156.173 g/mol
MDL Number: MFCD18074339
CAS Number: 392-61-0
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR