(R)-1-Cyclopropyl-2,2,2-trifluoroethan-1-amine hydrochloride ≥95%

Supplier: Ambeed
Warning

A507616-100MG A507616-250MG
77902-014EA 141 USD
77902-014 77901-031
(R)-1-Cyclopropyl-2,2,2-trifluoroethan-1-amine hydrochloride ≥95%
(R)-1-Cyclopropyl-2,2,2-trifluoroethan-1-amine hydrochloride

Precaución: for Research Use Only

Formula: C₅H₉ClF₃N
MDL Number: MFCD26575860
CAS Number: 1160756-75-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR