tert-Butyl 7-amino-2-azaspiro[4.4]nonane-2-carboxylate ≥98%

Supplier: Ambeed
Warning

A507580-250MG A507580-50MG
77617-168EA 301.79 USD
77617-168 77749-033
tert-Butyl 7-amino-2-azaspiro[4.4]nonane-2-carboxylate ≥98%
tert-Butyl 7-amino-2-azaspiro[4.4]nonane-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₃H₂₄N₂O₂
MDL Number: MFCD24384500
CAS Number: 1341037-08-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR