2-(3-(Bromomethyl)phenyl)acetic acid ≥95%

Supplier: Ambeed
Danger

A507185-1G A507185-25G
77884-279EA 74 USD
77884-279 77884-280
2-(3-(Bromomethyl)phenyl)acetic acid ≥95%
2-(3-(Bromomethyl)phenyl)acetic acid

Precaución: for Research Use Only

Formula: C₉H₉BrO₂
MDL Number: MFCD09842141
CAS Number: 118647-53-3
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR