(2R,3R,4R,5R)-2-(Acetoxymethyl)-5-(2-amino-8-bromo-6-oxo-1H-purin-9(6H)-yl)tetrahydrofuran-3,4-diyl diacetate ≥95%

Supplier: Ambeed
Warning

Synonyms: 2',3',5'-tri-O-acetyl-8-Bromoguanosine

A506952-1G A506952-250MG
77669-268EA 17 USD
77669-268 77872-843
(2R,3R,4R,5R)-2-(Acetoxymethyl)-5-(2-amino-8-bromo-6-oxo-1H-purin-9(6H)-yl)tetrahydrofuran-3,4-diyl diacetate ≥95%
(2R,3R,4R,5R)-2-(Acetoxymethyl)-5-(2-amino-8-bromo-6-oxo-1H-purin-9(6H)-yl)tetrahydrofuran-3,4-diyl diacetate

Precaución: for Research Use Only

Formula: C₁₆H₁₈BrN₅O₈
MDL Number: MFCD00057036
CAS Number: 15717-45-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR