N1-Benzylbenzene-1,2-diamine ≥95%

Supplier: Ambeed
Danger

A506882-1G A506882-250MG
77884-276EA 25 USD
77884-276 77884-277
N1-Benzylbenzene-1,2-diamine ≥95%
N1-Benzylbenzene-1,2-diamine

Precaución: for Research Use Only

Formula: C₁₃H₁₄N₂
MDL Number: MFCD03094682
CAS Number: 5822-13-9
UN: 3259
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR