2-Fluoro-(3-methoxycarbonyl)phenylboronic acid ≥98%, Ambeed.Inc

Supplier: Ambeed
Warning

Synonyms: 2-Fluoro-3-(methoxycarbonyl)benzeneboronic acid, 2-Fluoro-3-(methoxycarbonyl)phenylboronic acid, (2-Fluoro-3-(methoxycarbonyl)phenyl)boronic acid

A506432-100MG A506432-1G A506432-250MG A506432-5G
77684-328EA 33 USD
77684-328 77684-329 77886-300 77886-301
2-Fluoro-(3-methoxycarbonyl)phenylboronic acid ≥98%, Ambeed.Inc
2-Fluoro-(3-methoxycarbonyl)phenylboronic acid

Precaución: For research use only.

Formula: C₈H₈BFO₄
MW: 197.9561 g/mol
MDL Number: MFCD07374985
CAS Number: 1315476-07-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR