Boc-D-isoleucine ≥98%

Supplier: Ambeed
Warning

Synonyms: Boc-D-Ile-OH

A510574-100MG A510574-250MG A510574-5G
77882-256EA 50 USD
77882-256 77882-257 77882-258
Boc-D-isoleucine ≥98%
Boc-D-isoleucine

Precaución: for Research Use Only

Formula: C₁₁H₂₁NO₄
MW: 231.292 g/mol
MDL Number: MFCD00069989
CAS Number: 55721-65-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR