3,6-Dimethoxybenzene-1,2-diamine ≥97%

Supplier: Ambeed
Danger

A506418-5G A506418-100MG A506418-1G A506418-250MG
77594-588EA 198.85 USD
77594-588 77747-041 77747-042 77747-043
3,6-Dimethoxybenzene-1,2-diamine ≥97%
3,6-Dimethoxybenzene-1,2-diamine

Precaución: for Research Use Only

Formula: C₈H₁₂N₂O₂
MDL Number: MFCD20542271
CAS Number: 40328-95-8
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR