Cholesteryl chloride ≥98%

Supplier: Ambeed
Warning

A510458-100G A510458-25G A510458-5G
77602-186EA 244.98 USD
77602-186 77602-188 77602-190
Cholesteryl chloride ≥98%
Cholesteryl chloride

Precaución: for Research Use Only

Formula: C₂₇H₄₅Cl
MDL Number: MFCD00003632
CAS Number: 910-31-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR