1-(Toluene-4-sulfonyl)indole-3-boronic acid pinacol ester ≥98%

Supplier: Ambeed
Warning

Synonyms: 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1-tosyl-1H-indole

A510450-1G A510450-250MG A510450-5G
77886-306EA 86 USD
77886-306 77886-307 77886-308
1-(Toluene-4-sulfonyl)indole-3-boronic acid pinacol ester ≥98%
1-(Toluene-4-sulphonyl)indole-3-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₂₁H₂₄BNO₄S
MW: 397.3 g/mol
MDL Number: MFCD08063118
CAS Number: 1073354-51-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR