N-Cyclopropyl 3-boronobenzenesulfonamide ≥95%

Supplier: Ambeed
Warning

A510244-1G A510244-100MG A510244-250MG
77673-340EA 102 USD
77673-340 77884-282 77884-283
N-Cyclopropyl 3-boronobenzenesulfonamide ≥95%
N-Cyclopropyl 3-boronobenzenesulphonamide

Precaución: for Research Use Only

Formula: C₉H₁₂BNO₄S
MDL Number: MFCD08436035
CAS Number: 913835-28-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR