Imidazo[1,2-a]pyridine-6-carbaldehyde ≥96%

Supplier: Ambeed
Warning

A505378-1G A505378-250MG A505378-5G
77880-093EA 77 USD
77880-093 77880-094 77880-095
Imidazo[1,2-a]pyridine-6-carbaldehyde ≥96%
Imidazo[1,2-a]pyridine-6-carbaldehyde

Precaución: for Research Use Only

Formula: C₈H₆N₂O
MDL Number: MFCD07772808
CAS Number: 116355-16-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR