2-(2-(2-(2-Chloroethoxy)ethoxy)ethoxy)ethanol ≥95%

Supplier: Ambeed
Warning

A505358-1G A505358-250MG A505358-5G
77613-598EA 153.47 USD
77613-598 77673-324 77884-273
2-(2-(2-(2-Chloroethoxy)ethoxy)ethoxy)ethanol ≥95%
2-(2-(2-(2-Chloroethoxy)ethoxy)ethoxy)ethanol

Precaución: for Research Use Only

Formula: C₈H₁₇ClO₄
MDL Number: MFCD06252124
CAS Number: 5197-66-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR