tert-Butyl ((4-fluoropiperidin-4-yl)methyl)carbamate ≥98%, Ambeed.Inc

Supplier: Ambeed
Warning

Synonyms: tert-butyl N-[(4-fluoropiperidin-4-yl)methyl]carbamate

A505278-1G A505278-250MG A505278-5G
77613-592EA 126.74 USD
77613-592 77684-326 77893-014
tert-Butyl ((4-fluoropiperidin-4-yl)methyl)carbamate ≥98%, Ambeed.Inc
tert-Butyl ((4-fluoropiperidin-4-yl)methyl)carbamate

Precaución: For research use only.

Formula: C₁₁H₂₁FN₂O₂
MDL Number: MFCD11973738
CAS Number: 871022-62-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR