Fmoc-D-Phe(4-tBu)-OH ≥98%

Supplier: Ambeed
Warning

A509698-1G A509698-250MG A509698-5G
77901-120EA 35 USD
77901-120 77901-279 77901-260
Fmoc-D-Phe(4-tBu)-OH ≥98%
Fmoc-D-Phe(4-tBu)-OH

Precaución: for Research Use Only

Formula: C₂₈H₂₉NO₄
MDL Number: MFCD02094467
CAS Number: 252049-14-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR