4-(tert-Butoxy)benzotrifluoride ≥95%
Supplier: Ambeed
|
Warning
|
A509315-1G
A509315-5G
77686-327EA
15
USD
77686-327
77900-544
4-(tert-Butoxy)benzotrifluoride ≥95%
4-(tert-Butoxy)benzotrifluoride
Precaución: for Research Use Only
Formula:
C₁₁H₁₃F₃O |
MDL Number:
MFCD09763645 CAS Number: 16222-44-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...