5-Bromo-2-fluoro-4-methylbenzoic acid ≥98%

Supplier: Ambeed
Warning

A509287-250MG A509287-5G
77893-024EA 21 USD
77893-024 77893-025
5-Bromo-2-fluoro-4-methylbenzoic acid ≥98%
5-Bromo-2-fluoro-4-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆BrFO₂
MDL Number: MFCD23712067
CAS Number: 515135-65-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR