Fmoc-β-(1-piperazinyl)-Ala(Boc)-OH ≥98%

Supplier: Ambeed
Warning

Synonyms: (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(4-(tert-butoxycarbonyl)piperazin-1-yl)propanoic acid

A508740-1G A508740-250MG
77617-176EA 875.5 USD
77617-176 77747-048
Fmoc-β-(1-piperazinyl)-Ala(Boc)-OH ≥98%
Fmoc-β-(1-piperazinyl)-Ala(Boc)-OH

Precaución: for Research Use Only

Formula: C₂₇H₃₃N₃O₆
MDL Number: MFCD01860747
CAS Number: 313052-20-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR