5-((2,4-Diaminopyrimidin-5-yl)oxy)-4-isopropyl-2-methoxybenzenesulfonamide ≥98%

Supplier: Ambeed
Warning

Synonyms: Gefapixant

A503454-10MG A503454-1MG A503454-5MG
77598-528EA 123 USD
77598-528 77598-530 77598-532
5-((2,4-Diaminopyrimidin-5-yl)oxy)-4-isopropyl-2-methoxybenzenesulfonamide ≥98%
5-((2,4-Diaminopyrimidin-5-yl)oxy)-4-isopropyl-2-methoxybenzenesulfonamide

Precaución: for Research Use Only

Formula: C₁₄H₁₉N₅O₄S
MW: 353.4 g/mol
MDL Number: MFCD24539325
CAS Number: 1015787-98-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR