6-Bromopyridine-3-sulfonyl chloride ≥95%, Ambeed.Inc

Supplier: Ambeed
Danger

A491336-1G A491336-250MG A491336-100MG
77613-550EA 1239.09 USD
77613-550 77684-311 77891-000
6-Bromopyridine-3-sulfonyl chloride ≥95%, Ambeed.Inc
6-Bromopyridine-3-sulfonyl chloride

Precaución: For research use only.

Formula: C₅H₃BrClNO₂S
MDL Number: MFCD07374347
CAS Number: 886371-20-6
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR