Hexadec-15-ynoic acid ≥95%

Supplier: Ambeed
Warning

A491321-10MG A491321-1MG A491321-5MG
77686-281EA 214 USD
77686-281 77686-282 77686-283
Hexadec-15-ynoic acid ≥95%
Hexadec-15-ynoic acid

Precaución: for Research Use Only

Formula: C₁₆H₂₈O₂
MDL Number: MFCD22124928
CAS Number: 99208-90-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR