(R)-[1,1'-Binaphthalene]-2,2'-dicarboxylic acid ≥95%
Supplier: Ambeed
|
Warning
|
A503188-1G
A503188-250MG
77684-322EA
722
USD
77684-322
77684-323
(R)-[1,1'-Binaphthalene]-2,2'-dicarboxylic acid ≥95%
(R)-[1,1'-Binaphthalene]-2,2'-dicarboxylic acid
Precaución: For research use only.
Formula:
C₂₂H₁₄O₄ MW: 342.34 g/mol |
MDL Number:
MFCD00185729 CAS Number: 80703-23-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...