6-Undecanone ≥98%

Supplier: Ambeed
Warning

Synonyms: Dipentyl ketone, Diamyl ketone, Undecan-6-one

A491085-5G A491085-100G
77572-550EA 6.18 USD
77572-550 77594-534
6-Undecanone ≥98%
6-Undecanone

Precaución: for Research Use Only

Formula: C₁₁H₂₂O
MW: 170.3 g/mol
MDL Number: MFCD00009516
CAS Number: 927-49-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR