Astragaloside IV ≥98%, Ambeed.Inc

Supplier: Ambeed
Warning

A491011-10MG A491011-50MG A491011-1G A491011-250MG A491011-5G
77613-546EA 5.15 USD
77613-546 77684-309 77886-274 77886-275 77886-276
Astragaloside IV ≥98%, Ambeed.Inc
Astragaloside IV

Precaución: For research use only.

Formula: C₄₁H₆₈O₁₄
MDL Number: MFCD16036240
CAS Number: 84687-43-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR