2-Chloroquinolin-6-amine ≥95%

Supplier: Ambeed
Warning

A490744-100MG A490744-1G A490744-250MG
77684-306EA 109 USD
77684-306 77684-307 77684-308
2-Chloroquinolin-6-amine ≥95%
2-Chloroquinolin-6-amine

Precaución: For research use only.

Formula: C₉H₇ClN₂
MW: 178.61 g/mol
MDL Number: MFCD11847785
CAS Number: 238756-47-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR