2-Iodo-6-methoxybenzonitrile ≥98%

Supplier: Ambeed
Warning

A501935-1G A501935-250MG
77891-019EA 71 USD
77891-019 77891-020
2-Iodo-6-methoxybenzonitrile ≥98%
2-Iodo-6-methoxybenzonitrile

Precaución: for Research Use Only

Formula: C₈H₆INO
MDL Number: MFCD18905184
CAS Number: 66195-38-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR