2-Hydroxy-6-(trifluoromethyl)benzaldehyde ≥95%

Supplier: Ambeed
Warning

Synonyms: 6-(Trifluoromethyl)salicylaldehyde, 2-Formyl-3-hydroxybenzotrifluoride, 2-Formyl-3-(trifluoromethyl)phenol

A490218-1G A490218-250MG
77684-302EA 63 USD
77684-302 77684-303
2-Hydroxy-6-(trifluoromethyl)benzaldehyde ≥95%
2-Hydroxy-6-(trifluoromethyl)benzaldehyde

Precaución: For research use only.

Formula: C₈H₅F₃O₂
MW: 190.119 g/mol
MDL Number: MFCD16999701
CAS Number: 58914-35-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR