6-Methyl-1,3-dihydrofuro[3,4-c]pyridin-7-ol hydrochloride ≥95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Pyridoxine Impurity A
A501867-1G
A501867-250MG
A501867-100MG
A501867-5G
77598-524EA
311.6
USD
77598-524
77598-526
77617-142
77617-144
6-Methyl-1,3-dihydrofuro[3,4-c]pyridin-7-ol hydrochloride ≥95%
6-Methyl-1,3-dihydrofuro[3,4-c]pyridin-7-ol hydrochloride
Precaución: for Research Use Only
Formula:
C₈H₁₀ClNO₂ MW: 187.6235 g/mol |
MDL Number:
MFCD18803544 CAS Number: 1006-21-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...