1,2-Dipalmitoyl-sn-glycero-3-phosphoethanolamine ≥98%

Supplier: Ambeed
Warning

Synonyms: 1,2-Dipalmitoyl-sn-glycero-3-phosphoethanolamine, 1,2-Dihexadecanoyl-sn-glycero-3-phosphoethanolamine, Dipalmitoylphosphatidylethanolamine, 2-Aminoethyl (R)-2,3-Bis(hexadecanoyloxy)propyl Hydrogen Phosphate, (2R)-3-(((2-Aminoethoxy)(hydroxy)phosphoryl)oxy)propane-1,2-diyl dipalmitate, L-DPPE

A501616-1G A501616-250MG
77572-592EA 170.98 USD
77572-592 77686-314
1,2-Dipalmitoyl-sn-glycero-3-phosphoethanolamine ≥98%
1,2-Dipalmitoyl-sn-glycero-3-phosphoethanolamine

Precaución: for Research Use Only

Formula: C₃₇H₇₄NO₈P
MW: 691.97 g/mol
MDL Number: MFCD00036776
CAS Number: 923-61-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR