Methyl 2-(2-iodophenyl)acetate ≥95%
Supplier: Ambeed
|
Warning
|
A489784-5G
A489784-1G
77594-524EA
250.1
USD
77594-524
77684-300
Methyl 2-(2-iodophenyl)acetate ≥95%
Methyl 2-(2-iodophenyl)acetate
Precaución: For research use only.
Formula:
C₉H₉IO₂ MW: 276.07 g/mol |
MDL Number:
MFCD12025058 CAS Number: 66370-75-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...