2-((4-Chlorophenyl)amino)nicotinic acid ≥97%

Supplier: Ambeed
Warning

Synonyms: DHODH-IN-17

A501481-5MG A501481-10MG
77747-026EA 60 USD
77747-026 77747-025
2-((4-Chlorophenyl)amino)nicotinic acid ≥97%
2-((4-Chlorophenyl)amino)nicotinic acid

Precaución: for Research Use Only

Formula: C₁₂H₉ClN₂O₂
MW: 248.67 g/mol
MDL Number: MFCD03114306
CAS Number: 16344-26-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR