(S)-1-(5-Bromopyridin-2-yl)ethanamine ≥95%

Supplier: Ambeed
Warning

A501409-100MG A501409-250MG
77884-266EA 46 USD
77884-266 77884-267
(S)-1-(5-Bromopyridin-2-yl)ethanamine ≥95%
(S)-1-(5-Bromopyridin-2-yl)ethanamine

Precaución: for Research Use Only

Formula: C₇H₉BrN₂
MDL Number: MFCD09256735
CAS Number: 915720-70-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR