1-Bromo-2,4-dimethyl-3-nitrobenzene ≥97%

Supplier: Ambeed
Warning

A501347-25G A501347-5G A501347-1G A501347-250MG
77572-588EA 586.07 USD
77572-588 77613-580 77684-320 77684-321
1-Bromo-2,4-dimethyl-3-nitrobenzene ≥97%
1-Bromo-2,4-dimethyl-3-nitrobenzene

Precaución: For research use only.

Formula: C₈H₈BrNO₂
MW: 230.059 g/mol
MDL Number: MFCD00092653
CAS Number: 39053-43-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR