1-Boc-6-methylindole-2-boronic acid pinacol ester ≥98%

Supplier: Ambeed
Warning

Synonyms: 6-Methyl-1H-indole-2-boronic acid, N-BOC protected, (1-(tert-Butoxycarbonyl)-6-methyl-1H-indol-2-yl)boronic acid

A489599-250MG A489599-5G
77872-811EA 29 USD
77872-811 77872-812
1-Boc-6-methylindole-2-boronic acid pinacol ester ≥98%
1-Boc-6-methylindole-2-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₁₈BNO₄
MW: 275.11
MDL Number: MFCD04973738
CAS Number: 850568-51-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR