2-Chloro-6-methoxyphenol ≥97%

Supplier: Ambeed
Warning

A501095-100MG A501095-250MG
77884-264EA 28 USD
77884-264 77884-265
2-Chloro-6-methoxyphenol ≥97%
2-Chloro-6-methoxyphenol

Precaución: for Research Use Only

Formula: C₇H₇ClO₂
MDL Number: MFCD16998800
CAS Number: 72403-03-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR