2-Benzyloxy-1-bromonaphthalene 95+%

Supplier: Ambeed
Warning

A489053-1G A489053-250MG
77884-244EA 210 USD
77884-244 77884-245
2-Benzyloxy-1-bromonaphthalene 95+%
2-Benzyloxy-1-bromonaphthalene

Precaución: for Research Use Only

Formula: C₁₇H₁₃BrO
MDL Number: MFCD16295232
CAS Number: 41908-23-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR