3-Oxa-9-azaspiro[5.5]undecane hydrochloride ≥98%

Supplier: Ambeed
Warning

A500545-100MG A500545-1G A500545-250MG
77613-578EA 50.47 USD
77613-578 77891-016 77891-017
3-Oxa-9-azaspiro[5.5]undecane hydrochloride ≥98%
3-Oxa-9-azaspiro[5.5]undecane hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₈ClNO
MDL Number: MFCD16556308
CAS Number: 1380300-88-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR