3-Hydroxy-2-nitrobenzaldehyde ≥95%

Supplier: Ambeed
Warning

A500422-1G A500422-100MG A500422-250MG
77673-306EA 101 USD
77673-306 77884-260 77884-261
3-Hydroxy-2-nitrobenzaldehyde ≥95%
3-Hydroxy-2-nitrobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₅NO₄
MDL Number: MFCD09753744
CAS Number: 42123-33-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR