(6-((Tetrahydro-2H-pyran-4-yl)oxy)pyridin-3-yl)boronic acid ≥95%

Supplier: Ambeed
Warning

A500408-250MG A500408-5G
77886-289EA 116 USD
77886-289 77886-290
(6-((Tetrahydro-2H-pyran-4-yl)oxy)pyridin-3-yl)boronic acid ≥95%
(6-((Tetrahydro-2H-pyran-4-yl)oxy)pyridin-3-yl)boronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₄BNO₄
MDL Number: MFCD18261459
CAS Number: 1028745-48-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR