Methyl-3-methoxy-4-nitrobenzoate ≥98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 3-methoxy-4-nitrobenzoate, 3-Methoxy-4-nitrobenzoic acid methyl ester, Methyl 3-methoxy-4-nitrobenzenecarboxylate
A488519-10G
A488519-1G
A488519-5G
77880-062EA
115
USD
77880-062
77880-063
77880-064
Methyl-3-methoxy-4-nitrobenzoate ≥98%
Methyl-3-methoxy-4-nitrobenzoate
Precaución: for Research Use Only
Formula:
C₉H₉NO₅ |
MDL Number:
MFCD00051967 CAS Number: 5081-37-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...