Sodium glycocholate hydrate ≥95%, Ambeed.Inc
Supplier: Ambeed
|
Warning
|
A488277-1G
A488277-250MG
A488277-25G
A488277-5G
77684-297EA
30
USD
77684-297
77886-264
77886-265
77886-266
Sodium glycocholate hydrate ≥95%, Ambeed.Inc
Sodium glycocholate hydrate
Precaución: For research use only.
Formula:
C₂₆H₄₂NO₆Na·xH₂O |
MDL Number:
MFCD09037360 CAS Number: 207614-05-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...