1,8-Bis(maleimido)-3,6-dioxaoctane ≥98%

Supplier: Ambeed
Warning

A499662-250MG A499662-100MG
77747-023EA 27 USD
77747-023 77747-022
1,8-Bis(maleimido)-3,6-dioxaoctane ≥98%
1,8-Bis(maleimido)-3,6-dioxaoctane

Precaución: for Research Use Only

Formula: C₁₄H₁₆N₂O₆
MDL Number: MFCD01096749
CAS Number: 115597-84-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR