2-(5-Bromothiophen-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥96%

Supplier: Ambeed
Warning

A499562-1G A499562-250MG A499562-5G
77626-994EA 32.96 USD
77626-994 77878-274 77878-275
2-(5-Bromothiophen-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥96%
2-(5-Bromothiophen-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₀H₁₄BBrO₂S
MDL Number: MFCD08063136
CAS Number: 676501-84-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR