Bicyclo[2.2.1]heptane-1,4-dicarboxylic acid ≥97%, Ambeed.Inc
Supplier: Ambeed
|
Warning
|
A487935-1G
A487935-5G
A487935-250MG
A487935-25G
77594-518EA
117.88
USD
77594-518
77594-520
77684-295
77684-296
Bicyclo[2.2.1]heptane-1,4-dicarboxylic acid ≥97%, Ambeed.Inc
Bicyclo[2.2.1]heptane-1,4-dicarboxylic acid
Precaución: For research use only.
Formula:
C₉H₁₂O₄ MW: 184.19 g/mol |
MDL Number:
MFCD20639579 CAS Number: 15544-51-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...